Pipe blind lockouts
Pipe blind lockout, small (hand wheel diameter 13 - 76 mm), Red, 374,65×177,8×73,02 mm
N° de catalogue BRDY149228
Fournisseur: Brady
Jump to:
Product Details & Documents
These devices can be applied to the widest range of pipes to easily and temporarily avoid the release of gas, steam and hazardous liquids while pipe maintenance is still ongoing.
- Adjustable locking arm accommodates both standard and recessed bolt positions
- Secure 1 or 2 bolts for extra security
- Heavy duty construction withstands harsh environments
- UL approved for use in explosive atmospheres
Min. shackle clearance: 19,05 mm
Max. shackle diameter: 7,00 mm
Documents
Specifications
- Type:Pipe blind lockout, small (hand wheel diameter 13 - 76 mm)
- Colour:Red
- Seq:01
- W×D×H:374,65×177,8×73,02 mm



