Orders LinkContactUsLinkComponent
Tool carts
Tool carts
Tool cart, with 8 drawers (2 large, 2 medium and 4 small), secure lock and cleanroom castors with brakes, 900×600×1193 mm
Catalog # PALBTOOLCART-8-316
Supplier:   Palbam
Tool carts
Catalog # PALBTOOLCART-8-316
Supplier:  Palbam
Tool carts
Tool carts
Tool cart, with 8 drawers (2 large, 2 medium and 4 small), secure lock and cleanroom castors with brakes, 900×600×1193 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Tool cart in stainless steel 304 with electropolished finish.
  • Five fully extendable drawers
  • One lock which secures all the drawers
  • Top ledge to prevent tools/parts falling
  • Cleanroom castors with brakes
Specifications
  • Description:
    Tool cart, with 8 drawers (2 large, 2 medium and 4 small), secure lock and cleanroom castors with brakes
  • W×D×H:
    900×600×1193 mm