MULTIPURPOSE BEAKERS, PP, 200 ML, SCREW-CAP, STERILIZED 1 * 150 items
Catalog # RATI5921184
Supplier: RATIOLAB
MULTIPURPOSE BEAKERS, PP, 200 ML, SCREW-CAP, STERILIZED 1 * 150 items
Catalog # RATI5921184
Supplier: RATIOLAB
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Steam indicator tape, Comply™
Supplier: 3M MEDICAL
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Dialysis tubing, trial kits
Supplier: Spectrum Laboratories