Orders LinkContactUsLinkComponent
Blind cap
Blind cap
14,5/23 NS
Catalog # NEUB1670-10-14
Blind cap
Catalog # NEUB1670-10-14
Blind cap
14,5/23 NS
Price
19,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Ground cap with standard ground joints made of borosilicate glass 3.3.
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Cone size:
    14,5/23 NS
  • Seq:
    01