ContactUsLinkComponent
Fluorescence Polarization Filter with holder 535nm BW 25nm Polarized Vertical 1 * 1 items
Catalog # MLDV6590-0101
Supplier:  Molecular Devices
Fluorescence Polarization Filter with holder 535nm BW 25nm Polarized Vertical 1 * 1 items
Catalog # MLDV6590-0101
Supplier:  Molecular Devices
Frequently Bought Together