Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Trolleys, serving

Serving trolley, heavy duty, with reinforced chassis, SSW, 895×595×1015 mm, 18,33 kg, 2 (800×500 mm)
  HUPF0112172
Trolleys, serving
  HUPF0112172
Trolleys, serving
Trolleys, serving

Serving trolley, heavy duty, with reinforced chassis, SSW, 895×595×1015 mm, 18,33 kg, 2 (800×500 mm)

 
970,00 €
   
1 * 1 ST
 

Transport trolley complete with 25 mm deep shelves. Shelves complete with bevelled edges for user comfort. With four swivel castors, two of them with brake.

  • Corrosion resistant
  • Disc bumpers are made of elastic and shock resistant plastic material, it protect trolley from damage
  • Maintenance free

High-quality stainless steel construction.

 
  • Description:
    Serving trolley, heavy duty, with reinforced chassis, SSW
  • W×D×H:
    895×595×1015 mm
  • Shelves:
    2 (800×500 mm)
  • Weight:
    18,33 kg
Accessories

 

  •  
    Description
     
     
  • HUPF0114507
    Gallery, stainless steel, four sided, height 50 mm, for 600×400 mm shelves
       
    1 * 1 ST - 248,00 €

 

  •  
    Description
     
     
  • HUPF0114502
    Gallery, stainless steel, height: 50 mm, width: 799 mm, depth: 430 mm, for 800×500 mm shelves
       
    1 * 1 ST - 228,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.