Order Entry
Belgium
Orders LinkContactUsLinkComponent
DCpak® SFC Achiral (SFC- A) Columns

DCpak® SFC Achiral (SFC- A) Columns

SFC Achiral columns, Analytical, DCpak SFC-A, 5 µm, 2,1 mm, 250 mm
Catalog # DAICAA495
Supplier:   CHIRAL TECHNOLOGIES
DCpak® SFC Achiral (SFC- A) Columns
Catalog # DAICAA495
DCpak® SFC Achiral (SFC- A) Columns

SFC Achiral columns, Analytical, DCpak SFC-A, 5 µm, 2,1 mm, 250 mm

Price
642,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

When a material is in a state that the temperature and the pressure exceed the critical point, it becomes a supercritical fluid. In this phase, the density of gas and liquid becomes the same.

Specifications
  • Column Format:
    Analytical
  • Ø int.:
    2,1 mm
  • Particle Size:
    5 µm
  • Description:
    SFC Achiral columns
  • Length:
    250 mm
  • Seq:
    12
  • Phase:
    DCpak SFC-A
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.