ContactUsLinkComponent
General Purpose Transfer Pipettes, Large Bulb. Length 15,1 cm. Stem Diameter 6,1 mm. Overall Capacity 8,6 mL. Bulb Draw 4,7 mL. 21 drops per mL. 1 * 4.000 items
Catalog # SAMC204
Supplier:  SAMCO
General Purpose Transfer Pipettes, Large Bulb. Length 15,1 cm. Stem Diameter 6,1 mm. Overall Capacity 8,6 mL. Bulb Draw 4,7 mL. 21 drops per mL. 1 * 4.000 items
Catalog # SAMC204
Supplier:  SAMCO
Frequently Bought Together