[EN]INDIUM STANDARD SOLUTION 10000 G/ML 1 * 500 mL
Catalog # CHMBCL01.0923.0500
Supplier: CHEMLAB
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Latex Gloves
Supplier: VWR Collection