Order Entry
Belgium
Orders LinkContactUsLinkComponent
Erlenmeyer flasks with standard ground joints, amber

Erlenmeyer flasks with standard ground joints, amber

25 ml, Amber, 42 mm, 14/23 NS
Catalog # LENZ302808
Supplier:   Lenz Laborglas
Erlenmeyer flasks with standard ground joints, amber
Catalog # LENZ302808
Supplier:  Lenz Laborglas
Erlenmeyer flasks with standard ground joints, amber
Erlenmeyer flasks with standard ground joints, amber

25 ml, Amber, 42 mm, 14/23 NS

Price
21,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN® borosilicate glass, brown.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Coloured brown to protect light-sensitive substances. Volume graduations and labelling area.

Documents

Specifications
  • Ext.Ø:
    42 mm
  • Socket size:
    14/23 NS
  • Capacity:
    25 ml
  • Colour:
    Amber
  • Seq:
    03