Orders LinkContactUsLinkComponent
Robotic tips, Axygen®, for Beckman®
Robotic tips, Axygen®, for Beckman®
FX-384-XL-R (Extended), 0,5 - 30 µl, -, -, Transparent, AP-384, 50 racks, 384 each
Catalog # AXYGFX-384-XL-R
   Low Manufacturing Impact
Supplier:   Corning
Robotic tips, Axygen®, for Beckman®
Catalog # AXYGFX-384-XL-R
Supplier:  Corning
   Low Manufacturing Impact
Robotic tips, Axygen®, for Beckman®
Robotic tips, Axygen®, for Beckman®
FX-384-XL-R (Extended), 0,5 - 30 µl, -, -, Transparent, AP-384, 50 racks, 384 each
Price
4 820,00 €
Log in for availability
1 * 19.200 ST
Product Details & Documents
Free of RNase, DNase, DNA and pyrogens.
  • Transparent PP
  • Sterile tips: Sterilised by gamma radiation
  • Filter tips: PE filter prevents any contamination of the pipette
  • Maxymum Recovery™ tips: Ultra-smooth surface prevents retention, loss of components, and denaturation of the sample
Specifications
  • Packed:
    50 racks, 384 each
  • Sterile:
    -
  • Volume:
    0,5 - 30 µl
  • Tip style:
    FX-384-XL-R (Extended)
  • Tip colour:
    Transparent
  • For use with:
    AP-384
  • Filter:
    -
  • Cat. No.:
    AXYGFX-384-XL-R
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Low Manufacturing Impact