ContactUsLinkComponent
Spectratest FL1 fluorescence validation plate for MD fluorescence-capable microplate readers. Includes validation plate, calibration certificate, software protocols, manual, and storage case. 1 * 1 items
Catalog # MLDV0200-5060
Supplier:  Molecular Devices
Spectratest FL1 fluorescence validation plate for MD fluorescence-capable microplate readers. Includes validation plate, calibration certificate, software protocols, manual, and storage case. 1 * 1 items
Catalog # MLDV0200-5060
Supplier:  Molecular Devices
Frequently Bought Together