Orders LinkContactUsLinkComponent
Aluminium Sampling Case
Aluminium Sampling Case
Aluminium sampling case, 500 mm
Catalog # SAMP8509A-500
Supplier:   Sampling Systems
Aluminium Sampling Case
Catalog # SAMP8509A-500
Supplier:  Sampling Systems
Aluminium Sampling Case
Aluminium Sampling Case
Aluminium sampling case, 500 mm
Price
167,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
The aluminium sampling case is designed for storing samplers and equipment.
  • Available in a range of sizes
  • Lightweight and robust design
  • Foam lined to protect equipment when transported
  • Available from stock
  • Fast delivery
The sampling systems range of aluminium cases are perfect for any sampler storage requirements. They are made from robust and lightweight material and include an interior foam lining, ensuring that equipment is kept safe and protected at all times. The cases are lockable and come with a carrying handle.
Specifications
  • Description:
    Aluminium sampling case
  • Dimensions:
    500 mm
  • Seq:
    01