Orders LinkContactUsLinkComponent
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 40×/0,65/S (WD=0,6 mm), 0,6 mm
Catalog # MOTI1101001705231
Supplier:   MOTIC
Objectives, Panthera
Catalog # MOTI1101001705231
Supplier:  MOTIC
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 40×/0,65/S (WD=0,6 mm), 0,6 mm
Price
128,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    40x/0,65
  • Description:
    CCIS® Plan achromatic objective UC PL 40×/0,65/S (WD=0,6 mm)
  • Seq:
    05
  • Working distance:
    0,6 mm