Orders LinkContactUsLinkComponent
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 2×/0,05 (WD=7,2 mm) (To be used with swing-out condenser), 7,2 mm
Catalog # MOTI1101001705191
Supplier:   MOTIC
Objectives, Panthera
Catalog # MOTI1101001705191
Supplier:  MOTIC
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 2×/0,05 (WD=7,2 mm) (To be used with swing-out condenser), 7,2 mm
Price
321,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    2x/0,05
  • Description:
    CCIS® Plan achromatic objective UC PL 2×/0,05 (WD=7,2 mm) (To be used with swing-out condenser)
  • Seq:
    01
  • Working distance:
    7,2 mm