Orders LinkContactUsLinkComponent
96-Well plate sample control system, U-2D™
U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and chemical resistant non-slit mat
Catalog # MICS95296-500-1R
Supplier:   MICROSOLV
96-Well plate sample control system, U-2D™
Catalog # MICS95296-500-1R
Supplier:  MICROSOLV
96-Well plate sample control system, U-2D™
96-Well plate sample control system, U-2D™
U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and chemical resistant non-slit mat
Price
140,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The U-2D™ universal two dimensional 96-well plate system offer users unprecedented sample management control compared to current glass lined or plastic well plates for sample preparation or chromatography.
Easily push up the insert from the bottom and remove it from the rack. These inserts can then be placed into any MicroSolv 9 mm screw top autosampler vial for safe storage, transport or for a second chromatography dimension, or to another instrument platform.
 : Base and sealing mats sold separately.
Specifications
  • Description:
    U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and chemical resistant non-slit mat
  • Seq:
    21
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Paddles for U-2D™ sealing film
    Paddles for U-2D™ sealing film
    MICS95296-PDL-PK
    Log in for availability
    1 * 5 ST - 17,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Roller for U-2D™ sealing film
    Roller for U-2D™ sealing film
    MICS95296-ROLLER
    Log in for availability
    1 * 1 ST - 65,60 €
Product Information
  • Catalog #
    Availability
    Price
  • U-2D™ plate, 350µL, with rack and base only
    U-2D™ plate, 350µL, with rack and base only
    MICS95296-U2D
    Log in for availability
    1 * 1 ST - 116,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Zone free sealing film for U-2D™ plates
    Zone free sealing film for U-2D™ plates
    MICS95296-ZFF-PK
    Log in for availability
    1 * 10 ST - 54,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Zone free sealing film for U-2D™ plates
    Zone free sealing film for U-2D™ plates
    MICS95296-ZFF-CS
    Log in for availability
    1 * 50 ST - 228,00 €