ContactUsLinkComponent
Easy Purchase Pack. Autosampler vials and caps. 12x32mm screw top clear 1.2ml MRQ vials with silanized RSA glass and light blue AQR caps w/ clear silicone/PTFE septa in clear kit pack. AQ Brand. 100/pack. 1 * 100 items
Catalog # MICS9512S-1MP-DRS
Supplier:  MICROSOLV
undefined
Easy Purchase Pack. Autosampler vials and caps. 12x32mm screw top clear 1.2ml MRQ vials with silanized RSA glass and light blue AQR caps w/ clear silicone/PTFE septa in clear kit pack. AQ Brand. 100/pack. 1 * 100 items
Catalog # MICS9512S-1MP-DRS
Supplier:  MICROSOLV
Frequently Bought Together