Order Entry
Belgium
Orders LinkContactUsLinkComponent
Suite of columns, Cogent TYPE-C™, method development kits

Suite of columns, Cogent TYPE-C™, method development kits

Method development kit, 100 Å, 4 µm, 4,60 mm, 250 mm
Catalog # 554-6400
Supplier:   MICROSOLV
Suite of columns, Cogent TYPE-C™, method development kits
Catalog # 554-6400
Supplier:  MICROSOLV
Suite of columns, Cogent TYPE-C™, method development kits
Suite of columns, Cogent TYPE-C™, method development kits

Method development kit, 100 Å, 4 µm, 4,60 mm, 250 mm

Price
5 830,00 €
Log in for availability
1 * 5 ST
Product Details & Documents

The Cogent suite of columns is set of 5 columns of the same configuration consisting of one each of the Bidentate C18™, UDC-Cholesterol™, Bidentate C8™, Diamond Hydride™ and Silica C™.

  • Applicable in reverse phase, normal phase or aqueous normal phase
  • Kit provides an extremely wide spectrum of selectivies and possibilities

All of the columns in the suite can be used at low pH or high pH (upper limit is pH 8 for all except the C18 and the C8 columns which can be used up to pH 10,2).

All of the columns rapidly equilibrate between gradients or with initial use, making them very quick into service.

Specifications
  • Column Format:
    Method development kit
  • Ø int.:
    4,60 mm
  • Particle Size:
    4 µm
  • Pore size:
    100 Å
  • Length:
    250 mm
  • Seq:
    10
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.