ContactUsLinkComponent
CF680 with methyltetrazine reacts with cyclooctene (TCO, methylcyclopropene) labeled molecule via copper free click chemistry. This copper free bioorthogonal reaction allows staining the surface of live cells or having concerns about native protein f 1 * 1 mg
Catalog # 96039.
Supplier:  Biotium
CF680 with methyltetrazine reacts with cyclooctene (TCO, methylcyclopropene) labeled molecule via copper free click chemistry. This copper free bioorthogonal reaction allows staining the surface of live cells or having concerns about native protein f 1 * 1 mg
Catalog # 96039.
Supplier:  Biotium
Frequently Bought Together