Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH electrodes, Eutech

pH combination electrode, double junction, refillable, clog-free, 0 - 13, 0...+100 °C, Glass, Annular ceramic, 12×110 mm, BNC, 1 m cable, Samples containing TRIS buffers, sulfides and more
Catalog # EUTEECFG7370101B
Supplier:   EUTECH
pH electrodes, Eutech
Catalog # EUTEECFG7370101B
Supplier:  EUTECH
pH electrodes, Eutech
pH electrodes, Eutech

pH combination electrode, double junction, refillable, clog-free, 0 - 13, 0...+100 °C, Glass, Annular ceramic, 12×110 mm, BNC, 1 m cable, Samples containing TRIS buffers, sulfides and more

Price
297,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

pH electrodes for use with all pH meters with BNC input connector.

Specifications
  • Connection:
    BNC, 1 m cable
  • Ø×L:
    12×110 mm
  • For:
    Samples containing TRIS buffers, sulfides and more
  • Description:
    pH combination electrode, double junction, refillable, clog-free
  • Material:
    Glass
  • Seq:
    05
  • Diaphr.:
    Annular ceramic
  • Temp.:
    0...+100 °C
  • pH:
    0 - 13