ContactUsLinkComponent
Distilling links, with liebig condenser, w. thermometer socket st 14/23, flask and adapter cone st 14/23, jacket length 160 mm 1 * 1 items
Catalog # WITG1.100.160
Supplier:  witeg Labortechnik
Distilling links, with liebig condenser, w. thermometer socket st 14/23, flask and adapter cone st 14/23, jacket length 160 mm 1 * 1 items
Catalog # WITG1.100.160
Supplier:  witeg Labortechnik
Frequently Bought Together