ContactUsLinkComponent
RF313 WiFi Datalogger + 12-months FREE Cloud Application, large LCD, Integral Temperature and Humidity. Including wall bracket and micro USB Lead. -20°C to +60°C (-4°F to +140°F), 0 T0 100% rh. 1 * 1 items
Catalog # COMARF313-TH
Supplier:  COMARK
RF313 WiFi Datalogger + 12-months FREE Cloud Application, large LCD, Integral Temperature and Humidity. Including wall bracket and micro USB Lead. -20°C to +60°C (-4°F to +140°F), 0 T0 100% rh. 1 * 1 items
Catalog # COMARF313-TH
Supplier:  COMARK
Frequently Bought Together