Drying chambers
Tall clear acrylic chamber for bulk freeze drying, includes aluminium lid and 8 neoprene valves for use with both ¹/₂ and ³/₄" adapters, (requires shelves to operate) 442×584 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
Catalog # LBCP7318800
Supplier: LABCONCO
Jump to:
Product Details & Documents
Specifications
- Description:Tall clear acrylic chamber for bulk freeze drying, includes aluminium lid and 8 neoprene valves for use with both ¹/₂ and ³/₄" adapters, (requires shelves to operate) 442×584 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
- Seq:02
- Cat. No.:LBCP7318800
- Environmentally Preferable:Sustainable Packaging
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.