Finasteride 98%
Glass bottle, 5 g
Catalog # ACRO458000050
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
- CAS Number:98319-26-7
- Formula:C₂₃H₃₆N₂O₂
- MDL Number:MFCD00869737
- Molecular Weight:372.55 g/mol
- Melting Pt:252…254 °C
- Storage Temperature:Freezer
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Water:≤0.5 %
- HPLC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



