Finasteride 98%
Glass bottle, 1 g
ACRO458000010
: N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
- :98319-26-7
- :C₂₃H₃₆N₂O₂
- :MFCD00869737
- :372.55 g/mol
- :252…254 °C
- :Freezer
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Water:≤0.5 %
- HPLC:≥97.5 %
- Pack type:Glass bottle
- Pk:1 g



