Anthralin 97%
Glass bottle, 1 g
Catalog # ACRO457770010
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,8,9-Trihydroxyanthracene, 1,8,9-Anthracenetriol, DIT, Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-one
- CAS Number:1143-38-0
- Formula:C₁₄H₁₀O₃
- MDL Number:MFCD00053409
- Molecular Weight:226.23 g/mol
- Melting Pt:178…181 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:Yellow to red Crystalline powder
- Infrared spectrum:Conforms
- Melting point:178 to 182 °C
- HPLC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



