Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Urine containers, SpecTainer™ II
Urine containers, SpecTainer™ II
60 ml, Non sterile
Catalog # SIMPC567-60Y
Supplier:   Simport Scientific
Urine containers, SpecTainer™ II
Catalog # SIMPC567-60Y
Supplier:  Simport Scientific
Urine containers, SpecTainer™ II
Urine containers, SpecTainer™ II
60 ml, Non sterile
Price
438,00 €
Log in for availability
1 * 500 ST
Product Details & Documents
PP urine container with leak resistant PE screw cap, ideal for collecting, storing and transporting urine samples.
  • Graduated, except 60 ml container
  • Ridges around base offer a better grip during opening and closing
  • Stackable, shatter resistant
  • Designed with a straight side format
  • Moulded ridges around lid make it easy to open and close
Warning label has space for patient identification. For colour coding purposes, place a Capinsert™ on top of closure (see Series T345).
Specifications
  • Version:
    Non sterile
  • Capacité:
    60 ml
  • Seq:
    01