Orders LinkContactUsLinkComponent
Sight Boxes, PP
Sight Boxes, PP
Red, 495/460×31×185 mm
Catalog # SEMA8463
Supplier:   SEMADENI
Sight Boxes, PP
Catalog # SEMA8463
Supplier:  SEMADENI
Sight Boxes, PP
Red, 495/460×31×185 mm
Price
35,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of PP.
  • Cost-effective
  • Resistant to greases, resins, alkalis, oils and acids
  • Good temperature resistance
These storage boxes are for the professional sector and a wide range of applications in industry, crafts, household and hobby.
Specifications
  • Colour:
    Red
  • Seq:
    03
  • W×D×H:
    495/460×31×185 mm