Orders LinkContactUsLinkComponent
Microscopy scissors
Microscopy scissors
Blunt, straight, 125 mm
Catalog # RSGA101.125
Supplier:   RSG
Microscopy scissors
Catalog # RSGA101.125
Supplier:  RSG
Microscopy scissors
Microscopy scissors
Blunt, straight, 125 mm
Price
23,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Carbon steel, nickel plated, 8/10 mm.
Specifications
  • Type:
    Blunt, straight
  • Length:
    125 mm
  • Seq:
    02