Order Entry
Belgium
Orders LinkContactUsLinkComponent
Safe-Lock Tubes, 1,5 ml - Microtubes

Safe-Lock Tubes, 1,5 ml - Microtubes

1,5 ml, Colourless, Safe-Lock, PP, 22000×g, 500 tubes (10 bags × 50 tubes)
Catalog # 525-1042
   Low Manufacturing Impact
Supplier:   EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Catalog # 525-1042
Supplier:  EPPENDORF
   Low Manufacturing Impact
Safe-Lock Tubes, 1,5 ml - Microtubes
Safe-Lock Tubes, 1,5 ml - Microtubes

1,5 ml, Colourless, Safe-Lock, PP, 22000×g, 500 tubes (10 bags × 50 tubes)

Price
32,50 €
Log in for availability
1 * 500 ST
Product Details & Documents

Eppendorf® Safe-Lock 1,5 ml microtubes ensure secure sample storage. With a patented Safe-Lock design, these microtubes prevent accidental opening, safeguarding samples. Ideal for diverse applications, they offer reliability, durability, and ease of use in a 1,5 ml capacity.

  • Hinged Safe-Lock lid prevents inadvertent lid opening during incubation and storage
  • g-Safe® technology, depending on the Safe-Lock Tube variant, ensures outstanding centrifugation stability up to 30,000×g
  • g-Safe® technology prevents sample loss due to tube breakage, and provides additional safety when handling hazardous samples
  • Polypropylene construction offers robust resistance to chemicals, mechanical stress, and extreme temperatures
  • Precision lid sealing minimises evaporation rates during extended storage periods
  • Generously sized frosted lid and side surface facilitate easy labelling
  • Assured functionality in a temperature range from −86 to 100 °C
  • Autoclavable when open (121 °C, 20 min) for reliable sterilisation
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur® quality for tested purity

Eppendorf® Safe-Lock 1,5 ml microtubes exemplify secure sample storage with their patented Safe-Lock design, preventing accidental openings and preserving sample integrity. Versatile across laboratory applications, the durable construction and 1,5 ml capacity cater to varied sample volumes.

Clear graduations and a smooth surface enhance user-friendliness, ensuring a seamless and reliable laboratory experience. Elevate scientific endeavors with the trusted quality of Eppendorf® Safe-Lock Tubes.

Manufactured without the use of slip agents, plasticisers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA Grade can be downloaded from the Eppendorf website.

Specifications
  • Packed:
    500 tubes (10 bags × 50 tubes)
  • Type:
    Safe-Lock
  • Capacity:
    1,5 ml
  • Colour:
    Colourless
  • Material:
    PP
  • Seq:
    10
  • RCF max.:
    22000×g
  • Cat. No.:
    525-1042
  • Operating temperature range:
    −86...100 °C
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Volume:
    1,5 ml
  • Autoclavable:
    When open 121 °C, 20 min
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.