ContactUsLinkComponent
Burette 100:0.2ml CL.as with ns glass plug clear glass acc. to mohr,made of borosilicate glass 3.3 in compliance with din en iso 385 very good chemical resistance minimal thermal expansion high resistance to thermal shocks 1 * 2 items
Catalog # MARI2422041
Supplier:  MARIENFELD
Burette 100:0.2ml CL.as with ns glass plug clear glass acc. to mohr,made of borosilicate glass 3.3 in compliance with din en iso 385 very good chemical resistance minimal thermal expansion high resistance to thermal shocks 1 * 2 items
Catalog # MARI2422041
Supplier:  MARIENFELD
Frequently Bought Together