Order Entry
Belgium
Orders LinkContactUsLinkComponent
Retort rods, without thread

Retort rods, without thread

13 mm, 400 mm
Catalog # BOCH5180-400
Supplier:   BOCHEM
Retort rods, without thread
Catalog # BOCH5180-400
Supplier:  BOCHEM
Retort rods, without thread
Retort rods, without thread

13 mm, 400 mm

Price
26,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10.

  • Rods for stand bases
  • Without thread
Specifications
  • Ø:
    13 mm
  • Length:
    400 mm
  • Seq:
    09
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.