Funnel with short stem
100 mm, 10 mm, 100 mm
KAVAN632421001100
: Kavalier
Borosilicate glass, grade 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With smooth internal walls.
: DIN 12445 and ISO 4798
- Funnel O-Ø:100 mm
- Seq:14
- Stem O-Ø:10 mm
- Stem length:100 mm



