ContactUsLinkComponent
Thick-walled beaker made of PTFE, reinforced upper rim with spout. The base is made of heat-resistant PTFE-carbon, thus the beaker can be heated on a hot plate up to max. + 270 °C. The inside of the beaker is made of pure PTFE and provides universal 1 * 1 items
Catalog # BOHLA135-06
Supplier:  Bohlender
undefined
Thick-walled beaker made of PTFE, reinforced upper rim with spout. The base is made of heat-resistant PTFE-carbon, thus the beaker can be heated on a hot plate up to max. + 270 °C. The inside of the beaker is made of pure PTFE and provides universal 1 * 1 items
Catalog # BOHLA135-06
Supplier:  Bohlender
Frequently Bought Together