Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vessel cannulation forceps

Vessel cannulation forceps

0,35 mm Ext.Ø tubing size, Inox, 115 mm, Angled
Catalog # FSCI18403-11
Marketsource
Supplier:   FINE SCIENCE TOOLS
Vessel cannulation forceps
Catalog # FSCI18403-11
Supplier:  FINE SCIENCE TOOLS
Marketsource
Vessel cannulation forceps
Vessel cannulation forceps

0,35 mm Ext.Ø tubing size, Inox, 115 mm, Angled

Price
192,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    0,35 mm Ext.Ø tubing size
  • Length:
    115 mm
  • Material:
    Inox
  • Tip shape:
    Angled
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.