Orders LinkContactUsLinkComponent
Beer bottles
Beer bottles
-, With stainless steel wire
Catalog # RIXI8-1171-BM
Beer bottles
Catalog # RIXI8-1171-BM
Beer bottles
Beer bottles
-, With stainless steel wire
Price
1 440,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Glass, clear for clamp closure.
Ideal for transport and storage of beer samples.
Specifications
  • Type:
    With stainless steel wire
  • Capacity:
    -