ContactUsLinkComponent
Drop-n-Stain™ CF543 donkey anti-mouse IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting.  T o minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster 1 * 5 mL
Catalog # BTIU20967
Supplier:  Biotium
Drop-n-Stain™ CF543 donkey anti-mouse IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting.  T o minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster 1 * 5 mL
Catalog # BTIU20967
Supplier:  Biotium
Frequently Bought Together