Plate, staining ,12 place ,79 x 63 x 4 mm dimension,made of pressed glass,cavity 16mm, diameter 2mm depth 1 * 15 items
MARI1800003
Plate, staining ,12 place ,79 x 63 x 4 mm dimension,made of pressed glass,cavity 16mm, diameter 2mm depth 1 * 15 items
MARI1800003
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection