ContactUsLinkComponent
Mini temperature data logger for process monitoring. Tempmate B2 including 3-point calibration measuring range : - 40 C to +85C Calibration at -20C, +25C , +50C 1 * 1 items
Catalog # IMEC100521
Supplier:  tempmate
Mini temperature data logger for process monitoring. Tempmate B2 including 3-point calibration measuring range : - 40 C to +85C Calibration at -20C, +25C , +50C 1 * 1 items
Catalog # IMEC100521
Supplier:  tempmate
Frequently Bought Together