Funnel with short stem
60 mm, 8 mm, 60 mm
KAVAN632421001060
: Kavalier
Borosilicate glass, grade 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With smooth internal walls.
: DIN 12445 and ISO 4798
- Funnel O-Ø:60 mm
- Seq:07
- Stem O-Ø:8 mm
- Stem length:60 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



