Funnel with short stem
50 mm, 7 mm, 50 mm
Catalog # KAVAN632421001050
Supplier: Kavalier
Jump to:
Product Details & Documents
Borosilicate glass, grade 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With smooth internal walls.
: DIN 12445 and ISO 4798
Specifications
- Funnel O-Ø:50 mm
- Seq:05
- Stem O-Ø:7 mm
- Stem length:50 mm



