ContactUsLinkComponent
Blue adhering mat is an alternative to flask clamps or clamps for securing vessels to the Mini-300 Orbital-Genie platform. 11.8in x 11.8in (300mm x 300mm) Patented material is naturally adhering. 1 * 1 items
Catalog # SCIISI-M1617
undefined
Blue adhering mat is an alternative to flask clamps or clamps for securing vessels to the Mini-300 Orbital-Genie platform. 11.8in x 11.8in (300mm x 300mm) Patented material is naturally adhering. 1 * 1 items
Catalog # SCIISI-M1617
Frequently Bought Together