METHANOL LC/MS WITH 0.1% FORMIC ACID 1 * 2,5 L
Catalog # PRLS89782.320
Supplier: PROLABO CMP
Frequently Bought Together
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips, water hardness, Dosatest
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
Powder pillows, ammonia
Supplier: Hach