ContactUsLinkComponent
6 components 10mg/l each of Chloroform [CAS:67-66-3] 1,1 ,1-Trichloroethane [CAS:71-55-6] Trichloroethene [CAS:79-01-6] Tetrachloromethane [CAS:56-23-5] Tetrachloroethene [CAS:127-18-4] 1,2-Dichloroethane [CAS:107-06-2] in Methanol 1 * 1 ml
Catalog # CMPLF76D.10.ML.1.5
6 components 10mg/l each of Chloroform [CAS:67-66-3] 1,1 ,1-Trichloroethane [CAS:71-55-6] Trichloroethene [CAS:79-01-6] Tetrachloromethane [CAS:56-23-5] Tetrachloroethene [CAS:127-18-4] 1,2-Dichloroethane [CAS:107-06-2] in Methanol 1 * 1 ml
Catalog # CMPLF76D.10.ML.1.5
Frequently Bought Together