ContactUsLinkComponent
Tubing threaded 100x130x3.5mm gl90 ,with restriction,acc. to din 12216, with iso screw thread,made of borosilicate glass 1 * 1 items
Catalog # WITG0.183.090
Supplier:  witeg Labortechnik
Tubing threaded 100x130x3.5mm gl90 ,with restriction,acc. to din 12216, with iso screw thread,made of borosilicate glass 1 * 1 items
Catalog # WITG0.183.090
Supplier:  witeg Labortechnik
Frequently Bought Together