Orders LinkContactUsLinkComponent
BioSample cards
BioSample cards
Five circles card with perforated circles, also covers HIV and infectios diseases, 0,07 ml
Catalog # BINZ8.460.0012.A-N
Supplier:   Ahlstrom
BioSample cards
Catalog # BINZ8.460.0012.A-N
Supplier:  Ahlstrom
BioSample cards
BioSample cards
Five circles card with perforated circles, also covers HIV and infectios diseases, 0,07 ml
Price
108,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
These BioSample cards are designed for microvolume sampling, transport and ambient storage of biological samples. These cards are made of white and pure absorbent fibers and contain no wet-strength additives or chemicals. This feature ensures reproducible collection and minimise possible interferences for reliable analysis of targeted analytes like small molecules, proteins, nucleic acids or vitamins.
  • Reliable and reproducible sample collection
  • Even and uniform sample distribution
  • Customisable design and printing
  • Rigorous quality control
  • Each card has five/eight spots
General applications include protein-based analysis, lipid-based analysis, genomics, human identification and forensics. Indicated cards can also be used for HIV viral load testing and infectious disease screening (including Hepatitis B, C and Syphilis).
Certifications: Manufacturing according to ISO 13485:2016 and ISO 9001:2015 standards.
In Vitro Diagnostic medical device in compliance with the Directive 98/79/EC.
Ordering information: Drying racks are available to facilitate drying of the cards at ambient temperature.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Volume:
    0,07 ml
  • Description:
    Five circles card with perforated circles, also covers HIV and infectios diseases
  • Seq:
    05