Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multi-packs, Proline® Plus

Multi-packs, Proline® Plus

3-pack MAX, incl. three single channel pipettes (100 - 1000 µl, 500 - 5000 µl, 1000 - 10000 µl), Optifit Tip tray boxes (1× 10 - 1000 µl), Optifit Bulk-in-Bag tip packs (1× 5 ml, 1× 10 ml) and a linear stand
Catalog # 613-5739
   Low Manufacturing Impact
Supplier:   Sartorius
Multi-packs, Proline® Plus
Catalog # 613-5739
Supplier:  Sartorius
   Low Manufacturing Impact
Multi-packs, Proline® Plus
Multi-packs, Proline® Plus

3-pack MAX, incl. three single channel pipettes (100 - 1000 µl, 500 - 5000 µl, 1000 - 10000 µl), Optifit Tip tray boxes (1× 10 - 1000 µl), Optifit Bulk-in-Bag tip packs (1× 5 ml, 1× 10 ml) and a linear stand

Price
725,00 €
Log in for availability
1 * 1 Un.
Product Details & Documents

These Multipacks combine a set of Proline® Plus mechanical pipettes with matching tip trays and useful accessories. They offer an opportunity to test and get started with Proline® Plus mechanical pipettes.

  • Ergonomic design with light pipetting forces, comfortable handle and finger support for reduced risk of strain injuries
  • Minimised risk of contamination with Safe-Cone Filters and full autoclavability
  • Highly durable with strengthened structure also for heavier use

It is the widest range of pipettes for various users and applications.

Delivery information: All packs include a calibration tool that also acts as a tube opener, as well as the required instructions.

Specifications
  • Description:
    3-pack MAX, incl. three single channel pipettes (100 - 1000 µl, 500 - 5000 µl, 1000 - 10000 µl), Optifit Tip tray boxes (1× 10 - 1000 µl), Optifit Bulk-in-Bag tip packs (1× 5 ml, 1× 10 ml) and a linear stand
  • Seq:
    03
  • Cat. No.:
    613-5739
  • Environmental certification:
    Renewable energy certificate; ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.