Orders LinkContactUsLinkComponent
Multi-packs, Proline® Plus
Multi-packs, Proline® Plus
4-pack 100, incl. four single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 2× 0,5 - 200 µl, 1× 10 - 1000 µl) and a linear stand
Catalog # 613-5735
   Low Manufacturing Impact
Supplier:   Sartorius
Multi-packs, Proline® Plus
Catalog # 613-5735
Supplier:  Sartorius
   Low Manufacturing Impact
Multi-packs, Proline® Plus
Multi-packs, Proline® Plus
4-pack 100, incl. four single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 2× 0,5 - 200 µl, 1× 10 - 1000 µl) and a linear stand
Price
948,00 €
Log in for availability
1 * 1 Un.
Product Details & Documents
These Multipacks combine a set of Proline® Plus mechanical pipettes with matching tip trays and useful accessories. They offer an opportunity to test and get started with Proline® Plus mechanical pipettes.
  • Ergonomic design with light pipetting forces, comfortable handle and finger support for reduced risk of strain injuries
  • Minimised risk of contamination with Safe-Cone Filters and full autoclavability
  • Highly durable with strengthened structure also for heavier use
It is the widest range of pipettes for various users and applications.
Delivery information: All packs include a calibration tool that also acts as a tube opener, as well as the required instructions.
Specifications
  • Description:
    4-pack 100, incl. four single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 2× 0,5 - 200 µl, 1× 10 - 1000 µl) and a linear stand
  • Seq:
    06
  • Cat. No.:
    613-5735
  • Environmental certification:
    Renewable energy certificate; ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact