Orders LinkContactUsLinkComponent
Multipacks, Tacta®
Multipacks, Tacta®
4-pack 100, incl. four single channel pipettes (0,5 - 10 / 10 - 100 / 20 - 200 / 100 - 1000 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 10 - 1000 μl; 96 tips each) and one linear stand
Catalog # 613-5629
   Low Manufacturing Impact
Supplier:   Sartorius
Multipacks, Tacta®
Catalog # 613-5629
Supplier:  Sartorius
   Low Manufacturing Impact
Multipacks, Tacta®
Multipacks, Tacta®
4-pack 100, incl. four single channel pipettes (0,5 - 10 / 10 - 100 / 20 - 200 / 100 - 1000 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 10 - 1000 μl; 96 tips each) and one linear stand
Price
1 260,00 €
Log in for availability
1 * 1 Un.
Product Details & Documents
The Tacta® Multipacks offer an opportunity to test and get started with Tacta® mechanical pipettes. Tacta® mechanical pipette offers excellent performance and safety in manual pipetting. It is designed to maintain high accuracy and precision in repetitive and long-lasting pipetting series.
  • Reliable results, even over lengthy pipetting periods
  • Controlled and smooth tip ejection
  • The Optilock® system provides flexibility for volume adjustment and locking
There are different multipack options to choose from ranging from a set of 3 to a set of 5 pipettes.
Ordering information: All sets come with a linear stand for the safe storage of your pipettes as well as suitable racked or bulk-packed Optifit tips.
Specifications
  • Description:
    4-pack 100, incl. four single channel pipettes (0,5 - 10 / 10 - 100 / 20 - 200 / 100 - 1000 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 10 - 1000 μl; 96 tips each) and one linear stand
  • Seq:
    04
  • Cat. No.:
    613-5629
  • Environmental certification:
    Renewable energy certificate; ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact