Orders LinkContactUsLinkComponent
Universal cable lockouts, SafeLex
Universal cable lockouts, SafeLex
With 1 m cable
Catalog # BRDY145554
Supplier:   Brady
Universal cable lockouts, SafeLex
Catalog # BRDY145554
Supplier:  Brady
Universal cable lockouts, SafeLex
Universal cable lockouts, SafeLex
With 1 m cable
Price
53,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
The integrated hasp and cable in these cable lockouts provide a simple way to lock out multiple electrical or valve energy isolation points.
  • Lightweight construction for easy handling, carrying and operation
  • Device comes complete with vinyl coated steel lockout cable attachment
  • Allows up to six safety padlocks for a secure lockout
Specifications
  • Type:
    With 1 m cable
  • Seq:
    01